Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGGGAAAAGCTCACTTTAAACA[A/G]TTGGAACACCGGTGGCACTGTTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604679 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR7705 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR7705 - microRNA 7705 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PABPC1 - poly(A) binding protein cytoplasmic 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002568.3 | 3319 | Intron | NP_002559.2 | |||
XM_005250861.2 | 3319 | Missense Mutation | ACT,ATT | T,I 635 | XP_005250918.1 |