Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTCAGGCAAGAGTGCCCGCTAC[A/G]ACCGCTTCTCCGGGGGGCCCAGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605455 MIM: 606692 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB26 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB26 - RAB26, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNHG19 - small nucleolar RNA host gene 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD60 - small nucleolar RNA, C/D box 60 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAF7 - TNF receptor associated factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032271.2 | 147 | Missense Mutation | AAC,GAC | N,D 10 | NP_115647.2 | |
XM_005255627.4 | 147 | Missense Mutation | AAC,GAC | N,D 10 | XP_005255684.1 | |
XM_011522700.1 | 147 | UTR 5 | XP_011521002.1 |