Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTTAAATCGTTCCAGGTATTGA[A/G]GAAGCTTGGACTGCTCTGTTTCCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611989 | ||||||||||||||||||||
Literature Links: |
MRPS27 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPS27 - mitochondrial ribosomal protein S27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286748.1 | 987 | Missense Mutation | CCT,CTT | P,L 342 | NP_001273677.1 | |
NM_001286751.1 | 987 | Missense Mutation | CCT,CTT | P,L 272 | NP_001273680.1 | |
NM_015084.2 | 987 | Missense Mutation | CCT,CTT | P,L 328 | NP_055899.2 |