Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCTCTCGTGCCTTCAGGAGCGC[C/T]TGCCAACGGTACCGCTGTCCGGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612799 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EARS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EARS2 - glutamyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBFD1 - ubiquitin family domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019116.2 | 845 | Silent Mutation | CTG,TTG | L,L 213 | NP_061989.2 | |
XM_011545894.2 | 845 | Silent Mutation | CTG,TTG | L,L 213 | XP_011544196.1 | |
XM_017023459.1 | 845 | Silent Mutation | CTG,TTG | L,L 213 | XP_016878948.1 | |
XM_017023460.1 | 845 | Silent Mutation | CTG,TTG | L,L 213 | XP_016878949.1 |