Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCACCCCCAGGAATGGGGAGCGCT[G/T]GCTTTTCCGTGGGAAAATTCCACGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 111730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B4GALNT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B4GALNT2 - beta-1,4-N-acetyl-galactosaminyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159387.1 | 225 | Intron | NP_001152859.1 | |||
NM_001159388.1 | 225 | Intron | NP_001152860.1 | |||
NM_153446.2 | 225 | Missense Mutation | GGC,TGC | G,C 5 | NP_703147.2 | |
XM_011524314.2 | 225 | Missense Mutation | GGC,TGC | G,C 5 | XP_011522616.1 | |
XM_017024173.1 | 225 | Intron | XP_016879662.1 |