Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTGAAACAGGAGCCCGTCGGAG[A/G]AAGGGGCAGAAGGGAGTGGAGAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616024 | ||||||||||||||||||||
Literature Links: |
SCAF8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCAF8 - SR-related CTD associated factor 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286188.1 | 149 | Missense Mutation | GAA,GGA | E,G 35 | NP_001273117.1 | |
NM_001286189.1 | 149 | Intron | NP_001273118.1 | |||
NM_001286194.1 | 149 | Intron | NP_001273123.1 | |||
NM_001286199.1 | 149 | Intron | NP_001273128.1 | |||
NM_014892.4 | 149 | UTR 5 | NP_055707.3 |