Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGACTCAAAATATTTTTAATTCAA[C/T]AACAACCGCTGAAGAAAAGATTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4765 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4765 - microRNA 4765 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC27 - tetratricopeptide repeat domain 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193509.1 | 449 | Missense Mutation | ACA,ATA | T,I 9 | NP_001180438.1 | |
NM_017735.4 | 449 | Missense Mutation | ACA,ATA | T,I 59 | NP_060205.3 | |
XM_005264416.1 | 449 | Missense Mutation | ACA,ATA | T,I 59 | XP_005264473.1 | |
XM_011532958.1 | 449 | Missense Mutation | ACA,ATA | T,I 59 | XP_011531260.1 |