Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTGCCAACCTCCTTCTAGATAG[C/G]AAACGTCTTACAGATTGTGGAGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612399 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TLE6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TLE6 - transducin like enhancer of split 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143986.1 | 258 | Missense Mutation | GCA,GGA | A,G 76 | NP_001137458.1 | |
NM_024760.2 | 258 | UTR 5 | NP_079036.1 | |||
XM_005259645.2 | 258 | Missense Mutation | GCA,GGA | A,G 76 | XP_005259702.1 | |
XM_011528300.2 | 258 | Missense Mutation | GCA,GGA | A,G 76 | XP_011526602.1 | |
XM_011528301.1 | 258 | UTR 5 | XP_011526603.1 | |||
XM_011528306.1 | 258 | UTR 5 | XP_011526608.1 |