Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGTACGACACGGACCGCGACGGGC[A/G]TGTGGGTTGGGAGGAGCTGCGCAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601437 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCGRT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FCGRT - Fc fragment of IgG receptor and transporter | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RCN3 - reticulocalbin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020650.2 | 1116 | Missense Mutation | CAT,CGT | H,R 132 | NP_065701.2 | |
XM_005259089.3 | 1116 | Intron | XP_005259146.1 | |||
XM_011527143.1 | 1116 | Missense Mutation | CAT,CGT | H,R 132 | XP_011525445.1 | |
XM_017027023.1 | 1116 | Missense Mutation | CAT,CGT | H,R 132 | XP_016882512.1 |