Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTGTAAAGCCTGGTGCCCATCT[A/G]ACAGTGAAGAAAATTTTTGTTGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605372 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HNRNPA3 - heterogeneous nuclear ribonucleoprotein A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005246380.1 | 475 | Silent Mutation | CTA,CTG | L,L 123 | XP_005246437.1 | |
XM_005246381.4 | 475 | Silent Mutation | CTA,CTG | L,L 101 | XP_005246438.1 | |
XM_006712365.2 | 475 | Silent Mutation | CTA,CTG | L,L 123 | XP_006712428.1 | |
XM_017003587.1 | 475 | Silent Mutation | CTA,CTG | L,L 101 | XP_016859076.1 | |
XM_017003588.1 | 475 | Silent Mutation | CTA,CTG | L,L 123 | XP_016859077.1 |
LOC105373759 - uncharacterized LOC105373759 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4444-1 - microRNA 4444-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |