Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCGCAGCCAGAGGCGCAGCGTGAG[C/T]TCGACGTGGTTCTCAGAACCGTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603402 MIM: 614309 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACOX3 - acyl-CoA oxidase 3, pristanoyl | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101667.1 | 582 | Intron | NP_001095137.1 | |||
NM_003501.2 | 582 | Intron | NP_003492.2 | |||
XM_005248011.4 | 582 | Intron | XP_005248068.1 | |||
XM_005248013.3 | 582 | Intron | XP_005248070.1 | |||
XM_011513565.2 | 582 | Intron | XP_011511867.1 | |||
XM_011513566.1 | 582 | Intron | XP_011511868.1 |
TRMT44 - tRNA methyltransferase 44 homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152544.2 | 582 | Missense Mutation | CTC,TTC | L,F 179 | NP_689757.2 | |
XM_005247950.3 | 582 | UTR 5 | XP_005248007.1 | |||
XM_011513406.2 | 582 | Missense Mutation | CTC,TTC | L,F 179 | XP_011511708.1 | |
XM_011513407.2 | 582 | Missense Mutation | CTC,TTC | L,F 179 | XP_011511709.1 | |
XM_011513408.1 | 582 | UTR 5 | XP_011511710.1 | |||
XM_011513409.2 | 582 | UTR 5 | XP_011511711.1 | |||
XM_011513410.2 | 582 | Missense Mutation | CTC,TTC | L,F 179 | XP_011511712.1 | |
XM_017007811.1 | 582 | Missense Mutation | CTC,TTC | L,F 179 | XP_016863300.1 |