Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATTCTAGGAGTCGAACTTCCAAC[A/G]GTATATGCATTTTGATATTACGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616190 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CMTR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CMTR2 - cap methyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099642.1 | 2337 | Missense Mutation | CCG,CTG | P,L 596 | NP_001093112.1 | |
NM_001324374.1 | 2337 | Missense Mutation | CCG,CTG | P,L 596 | NP_001311303.1 | |
NM_001324377.1 | 2337 | Missense Mutation | CCG,CTG | P,L 596 | NP_001311306.1 | |
NM_001324378.1 | 2337 | Missense Mutation | CCG,CTG | P,L 596 | NP_001311307.1 | |
NM_001324379.1 | 2337 | Missense Mutation | CCG,CTG | P,L 596 | NP_001311308.1 | |
NM_018348.5 | 2337 | Missense Mutation | CCG,CTG | P,L 596 | NP_060818.4 | |
XM_011523228.1 | 2337 | Missense Mutation | CCG,CTG | P,L 596 | XP_011521530.1 | |
XM_011523229.1 | 2337 | Missense Mutation | CCG,CTG | P,L 596 | XP_011521531.1 | |
XM_017023444.1 | 2337 | Missense Mutation | CCG,CTG | P,L 596 | XP_016878933.1 |