Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAACATCCCCAGCTAAGATAGTC[A/G]CGATTTCGCCCTGCATGAAGGCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611720 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRF2BPL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IRF2BPL - interferon regulatory factor 2 binding protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024496.3 | 3249 | Missense Mutation | GCG,GTG | A,V 781 | NP_078772.1 |
LOC105370579 - uncharacterized LOC105370579 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107984638 - uncharacterized LOC107984638 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |