Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTACGCAGATCCCATAAAGCTAC[A/G]GTCTAAAGAAAAATAACAAGTGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300825 MIM: 300677 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBBP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBBP7 - RB binding protein 7, chromatin remodeling factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198719.1 | 1246 | Silent Mutation | ACC,ACT | T,T 340 | NP_001185648.1 | |
NM_002893.3 | 1246 | Silent Mutation | ACC,ACT | T,T 296 | NP_002884.1 |
TXLNG - taxilin gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |