Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGTCATTGCTGATAAAAATGGCA[C/T]GGACTTCTATTCGTCTGTCCGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRINP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRINP3 - BMP/retinoic acid inducible neural specific 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317188.1 | 2107 | Missense Mutation | CAT,CGT | H,R 409 | NP_001304117.1 | |
NM_199051.2 | 2107 | Missense Mutation | CAT,CGT | H,R 511 | NP_950252.1 | |
XM_011509472.2 | 2107 | Missense Mutation | CAT,CGT | H,R 481 | XP_011507774.1 | |
XM_011509475.2 | 2107 | Missense Mutation | CAT,CGT | H,R 468 | XP_011507777.1 | |
XM_011509476.2 | 2107 | Missense Mutation | CAT,CGT | H,R 409 | XP_011507778.1 | |
XM_017001125.1 | 2107 | Missense Mutation | CAT,CGT | H,R 511 | XP_016856614.1 | |
XM_017001126.1 | 2107 | Missense Mutation | CAT,CGT | H,R 511 | XP_016856615.1 | |
XM_017001127.1 | 2107 | Missense Mutation | CAT,CGT | H,R 481 | XP_016856616.1 | |
XM_017001128.1 | 2107 | Missense Mutation | CAT,CGT | H,R 481 | XP_016856617.1 | |
XM_017001129.1 | 2107 | Missense Mutation | CAT,CGT | H,R 409 | XP_016856618.1 | |
XM_017001130.1 | 2107 | Missense Mutation | CAT,CGT | H,R 409 | XP_016856619.1 | |
XM_017001131.1 | 2107 | Missense Mutation | CAT,CGT | H,R 218 | XP_016856620.1 | |
XM_017001132.1 | 2107 | Intron | XP_016856621.1 | |||
XM_017001133.1 | 2107 | Intron | XP_016856622.1 |