Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTGTGCTTGATGACACCAACCA[C/T]GAACGGGAACGGCTGGAGCAGCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123830 MIM: 601964 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNP - 2',3'-cyclic nucleotide 3' phosphodiesterase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033133.4 | 546 | Silent Mutation | CAC,CAT | H,H 126 | NP_149124.3 | |
XM_006721701.3 | 546 | Silent Mutation | CAC,CAT | H,H 106 | XP_006721764.1 | |
XM_011524340.2 | 546 | Silent Mutation | CAC,CAT | H,H 106 | XP_011522642.1 |
DNAJC7 - DnaJ heat shock protein family (Hsp40) member C7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC25 - tetratricopeptide repeat domain 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |