Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGCTGAGTGATCGATAGATTCTG[C/T]GTACAGTATCACGCCTTCTGAATCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 176880 | |||||||||||||||||||||||
Literature Links: |
PROS1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PROS1 - protein S (alpha) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000313.3 | 1449 | Missense Mutation | ACA,GCA | T,A 341 | NP_000304.2 | |
NM_001314077.1 | 1449 | Missense Mutation | ACA,GCA | T,A 373 | NP_001301006.1 |