Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACATTGCTAACCACACAGACATA[A/G]GCCCCTCTGTCCCGTAGCTGCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609794 MIM: 602943 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINGO4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINGO4 - leucine rich repeat and Ig domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004432.3 | 1669 | Silent Mutation | GCC,GCT | A,A 481 | NP_001004432.1 |
RORC - RAR related orphan receptor C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |