Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGTGCTGGGGCGCGGCGCACGCG[C/T]GTCATCTACGTGCACAACAGCCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600222 | ||||||||||||||||||||
Literature Links: |
TIE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TIE1 - tyrosine kinase with immunoglobulin like and EGF like domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253357.1 | 439 | Silent Mutation | CGC,CGT | R,R 70 | NP_001240286.1 | |
NM_005424.4 | 439 | Silent Mutation | CGC,CGT | R,R 115 | NP_005415.1 | |
XM_005271163.1 | 439 | Silent Mutation | CGC,CGT | R,R 115 | XP_005271220.1 | |
XM_006710869.1 | 439 | UTR 5 | XP_006710932.1 | |||
XM_017002207.1 | 439 | Silent Mutation | CGC,CGT | R,R 115 | XP_016857696.1 |