Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGGTGCCTCTTCATGTGCAGGG[C/T]CAGGTGGTCAGACCTGGAAAAACAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602053 | ||||||||||||||||||||
Literature Links: |
KLF6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLF6 - Kruppel like factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160124.1 | 969 | Missense Mutation | ACC,GCC | T,A 234 | NP_001153596.1 | |
NM_001160125.1 | 969 | Nonsense Mutation | TGA,TGG | *,W 234 | NP_001153597.1 | |
NM_001300.5 | 969 | Missense Mutation | ACC,GCC | T,A 276 | NP_001291.3 |