Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACGCGGCGGTGACGACTTAGTCC[A/G]GAAGCATCACAGTAGGTCTTGTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159465 MIM: 612192 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MOG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MOG - myelin oligodendrocyte glycoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZFP57 - ZFP57 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109809.2 | 698 | Silent Mutation | TCC,TCT | S,S 244 | NP_001103279.2 | |
XM_006715087.3 | 698 | Silent Mutation | TCC,TCT | S,S 172 | XP_006715150.1 | |
XM_011514570.2 | 698 | Silent Mutation | TCC,TCT | S,S 244 | XP_011512872.1 |