Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTGAATGTGAGCAACTACAGCC[A/G]TGTGTCTCTGCAGAATGCCACCTCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607952 | |||||||||||||||||||||||
Literature Links: |
SLC6A11 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SLC6A11 - solute carrier family 6 member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317406.1 | 644 | Missense Mutation | CAT,CGT | H,R 193 | NP_001304335.1 | |
NM_014229.2 | 644 | Missense Mutation | CAT,CGT | H,R 193 | NP_055044.1 | |
XM_011534033.2 | 644 | Missense Mutation | CAT,CGT | H,R 193 | XP_011532335.1 | |
XM_017007073.1 | 644 | Missense Mutation | ATG,GTG | M,V 1 | XP_016862562.1 |