Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCTGCGGGGGCTCGGCCTGCTC[C/T]TGCCCGGGCTCCGCATGGGTAGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZSCAN10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZSCAN10 - zinc finger and SCAN domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282415.1 | 2078 | Silent Mutation | CAA,CAG | Q,Q 323 | NP_001269344.1 | |
NM_001282416.1 | 2078 | Silent Mutation | CAA,CAG | Q,Q 580 | NP_001269345.1 | |
NM_032805.2 | 2078 | Silent Mutation | CAA,CAG | Q,Q 717 | NP_116194.2 | |
XM_017023791.1 | 2078 | Silent Mutation | CAA,CAG | Q,Q 717 | XP_016879280.1 | |
XM_017023792.1 | 2078 | Silent Mutation | CAA,CAG | Q,Q 685 | XP_016879281.1 |