Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGAAACTCCACAAATGCCAGTC[A/G]GTGCTCCCAGTGCCCTCGACCCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176894 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRKG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRKG1 - protein kinase, cGMP-dependent, type I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098512.2 | 363 | Silent Mutation | TCA,TCG | S,S 45 | NP_001091982.1 | |
NM_006258.3 | 363 | Intron | NP_006249.1 | |||
XM_011539952.2 | 363 | Intron | XP_011538254.1 | |||
XM_017016412.1 | 363 | Intron | XP_016871901.1 | |||
XM_017016413.1 | 363 | Intron | XP_016871902.1 |