Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATCTTGGAATTGAAGTAATTCGC[C/T]TGTTTTTTGGTAAGTGTTGTCCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613277 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPSF7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPSF7 - cleavage and polyadenylation specific factor 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM216 - transmembrane protein 216 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173990.2 | 1179 | Silent Mutation | CTG,TTG | L,L 74 | NP_001167461.1 | |
NM_001173991.2 | 1179 | Silent Mutation | CTG,TTG | L,L 74 | NP_001167462.1 | |
NM_016499.5 | 1179 | Silent Mutation | CTG,TTG | L,L 13 | NP_057583.2 | |
XM_005274039.3 | 1179 | Silent Mutation | CTG,TTG | L,L 13 | XP_005274096.1 | |
XM_017017882.1 | 1179 | Silent Mutation | CTG,TTG | L,L 13 | XP_016873371.1 |