Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAGACTCGCTCAGGCACTTTGGG[A/C]GACGTGAAATACGCGGGGGTCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603244 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HS3ST1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HS3ST1 - heparan sulfate-glucosamine 3-sulfotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005114.2 | 5460 | Silent Mutation | NP_005105.1 | |||
XM_011513913.2 | 5460 | Silent Mutation | XP_011512215.1 |