Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTATCTATCACTTACCCTCCTGG[G/T]TAGCTTTGTGTACCGAACATAATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610657 MIM: 602019 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0196 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0196 - KIAA0196 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014846.3 | 3949 | Missense Mutation | ACC,CCC | T,P 1139 | NP_055661.3 | |
XM_005251120.3 | 3949 | Missense Mutation | ACC,CCC | T,P 991 | XP_005251177.1 | |
XM_011517409.1 | 3949 | Intron | XP_011515711.1 | |||
XM_017014113.1 | 3949 | Missense Mutation | ACC,CCC | T,P 1139 | XP_016869602.1 |
SQLE - squalene epoxidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |