Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGATGCCCTCAGAAGATGCCCTT[A/G]GCAATCTTGAGTCCTTTCTGCTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601619 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RALGDS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RALGDS - ral guanine nucleotide dissociation stimulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042368.2 | 2810 | Silent Mutation | GCC,GCT | A,A 855 | NP_001035827.1 | |
NM_001271774.1 | 2810 | Silent Mutation | GCC,GCT | A,A 881 | NP_001258703.1 | |
NM_001271775.1 | 2810 | Silent Mutation | GCC,GCT | A,A 909 | NP_001258704.1 | |
NM_001271776.1 | 2810 | Silent Mutation | GCC,GCT | A,A 898 | NP_001258705.1 | |
NM_006266.3 | 2810 | Silent Mutation | GCC,GCT | A,A 910 | NP_006257.1 |