Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAATACCACATCTCCATTTCCTTC[C/T]GGTGGCTTGGGATGTCAGCATTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFT22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFT22 - intraflagellar transport 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130820.2 | 474 | Missense Mutation | CAG,CGG | Q,R 66 | NP_001124292.1 | |
NM_001130821.2 | 474 | Missense Mutation | CAG,CGG | Q,R 19 | NP_001124293.1 | |
NM_001130822.2 | 474 | Missense Mutation | CAG,CGG | Q,R 19 | NP_001124294.1 | |
NM_001287525.1 | 474 | Missense Mutation | CAG,CGG | Q,R 19 | NP_001274454.1 | |
NM_001287526.1 | 474 | Missense Mutation | CAG,CGG | Q,R 19 | NP_001274455.1 | |
NM_022777.3 | 474 | Missense Mutation | CAG,CGG | Q,R 96 | NP_073614.1 |
LOC101927746 - uncharacterized LOC101927746 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |