Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGATAATGTCCGGCCTCTGGCCT[A/G]TCCTGATTCTGATGCTGTGCTCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601555 MIM: 604631 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RND2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RND2 - Rho family GTPase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005440.4 | 174 | Missense Mutation | TAT,TGT | Y,C 76 | NP_005431.1 | |
XM_011525316.1 | 174 | Missense Mutation | TAT,TGT | Y,C 76 | XP_011523618.1 | |
XM_011525317.2 | 174 | Missense Mutation | TAT,TGT | Y,C 48 | XP_011523619.1 |
VAT1 - vesicle amine transport 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |