Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGGTGGTGGCAGATGAGGGTGA[C/T]TTTCAATGGTCACTGTTTTCTTCAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609606 | |||||||||||||||||||||||
Literature Links: |
ATG3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ATG3 - autophagy related 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278712.1 | 791 | Missense Mutation | AAT,AGT | N,S 249 | NP_001265641.1 | |
NM_022488.4 | 791 | Missense Mutation | AAT,AGT | N,S 249 | NP_071933.2 | |
XM_011513074.1 | 791 | Missense Mutation | AAT,AGT | N,S 162 | XP_011511376.1 |