Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATCTCTAACAAATCTTTCTTTG[A/G]GGATCTGTGTTTTAATTCAGAATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612758 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAPT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAPT1 - transmembrane anterior posterior transformation 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153365.2 | 2825 | Missense Mutation | CCA,TCA | P,S 549 | NP_699196.2 | |
XM_005248139.2 | 2825 | Missense Mutation | CCA,TCA | P,S 423 | XP_005248196.1 | |
XM_005248140.3 | 2825 | Missense Mutation | CCA,TCA | P,S 423 | XP_005248197.1 | |
XM_011513812.2 | 2825 | Missense Mutation | CCA,TCA | P,S 504 | XP_011512114.1 | |
XM_011513815.2 | 2825 | Missense Mutation | CCA,TCA | P,S 423 | XP_011512117.1 | |
XM_011513816.2 | 2825 | Missense Mutation | CCA,TCA | P,S 438 | XP_011512118.1 | |
XM_011513817.2 | 2825 | Missense Mutation | CCA,TCA | P,S 306 | XP_011512119.1 | |
XM_017007876.1 | 2825 | Missense Mutation | CCA,TCA | P,S 408 | XP_016863365.1 |