Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACGGGGCTCAGCTTCAGAGGGAC[C/G]AGTAGGGCCATTGTTGACATTGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300460 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCDH19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCDH19 - protocadherin 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105243.1 | 4851 | Missense Mutation | CGT,GGT | R,G 1060 | NP_001098713.1 | |
NM_001184880.1 | 4851 | Missense Mutation | CGT,GGT | R,G 1107 | NP_001171809.1 | |
NM_020766.2 | 4851 | Missense Mutation | CGT,GGT | R,G 1059 | NP_065817.2 | |
XM_011530997.2 | 4851 | Missense Mutation | CGT,GGT | R,G 1106 | XP_011529299.1 |