Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCGGTGGCTCCTCCTGGCCCAGC[A/C]GCAGAGGCGGTTTGCATTTGTGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612234 MIM: 612235 MIM: 612333 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALHM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CALHM1 - calcium homeostasis modulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001412.3 | 1081 | Missense Mutation | NP_001001412.3 |
CALHM2 - calcium homeostasis modulator 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDCD11 - programmed cell death 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |