Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAATTCAGGACGTTGTACCGTGT[A/G]ATGTGAACCAGCTTCTCAGCTCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300108 MIM: 300767 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIAPH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DIAPH2 - diaphanous related formin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006729.4 | 551 | Intron | NP_006720.1 | |||
NM_007309.3 | 551 | Intron | NP_009293.1 |
RPA4 - replication protein A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013347.4 | 551 | Missense Mutation | AAT,GAT | N,D 50 | NP_037479.1 |