Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTGTTCTCCCGGGGGTTGTAGC[A/G]CAGCCCGTGCGGCCCCTCGGAGTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613378 MIM: 232000 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGACT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GGACT - gamma-glutamylamine cyclotransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195087.1 | 787 | Missense Mutation | CGC,TGC | R,C 146 | NP_001182016.1 | |
NM_033110.2 | 787 | Missense Mutation | CGC,TGC | R,C 146 | NP_149101.1 | |
XM_005254083.2 | 787 | Missense Mutation | CGC,TGC | R,C 146 | XP_005254140.1 | |
XM_011521129.2 | 787 | Missense Mutation | CGC,TGC | R,C 146 | XP_011519431.1 | |
XM_017020804.1 | 787 | Missense Mutation | CGC,TGC | R,C 146 | XP_016876293.1 |
PCCA - propionyl-CoA carboxylase alpha subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |