Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACAGGCCTGTCCTGCGTCTACC[G/T]GCTCCACGCAGCTAGTGAGGCTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614911 MIM: 603846 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1QTNF4 - C1q and tumor necrosis factor related protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM180B - family with sequence similarity 180 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164379.1 | 483 | Missense Mutation | CGG,CTG | R,L 131 | NP_001157851.1 |
KBTBD4 - kelch repeat and BTB domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFS3 - NADH:ubiquinone oxidoreductase core subunit S3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |