Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCAGCTCCCCTGGCTCAGGGAC[A/G]GAGGCGTGTACAGTGCCCACCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608077 MIM: 603752 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR649 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR649 - microRNA 649 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
P2RX6 - purinergic receptor P2X 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC7A4 - solute carrier family 7 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004173.2 | 1421 | Silent Mutation | TCC,TCT | S,S 451 | NP_004164.2 |