Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTAAATAACGTCTCAGGAGGCCG[G/C]AGCCACGCTAGATGGGCTCCGGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612600 MIM: 612895 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRM3 - mitochondrial rRNA methyltransferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NXN - nucleoredoxin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205319.1 | 1222 | UTR 3 | NP_001192248.1 | |||
NM_022463.4 | 1222 | UTR 3 | NP_071908.2 | |||
XM_005256756.4 | 1222 | UTR 3 | XP_005256813.1 | |||
XM_005256758.3 | 1222 | UTR 3 | XP_005256815.1 | |||
XM_017024949.1 | 1222 | Intron | XP_016880438.1 |