Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGATCGAGAGGAGCCGGCAGCT[A/G]CAGAAGGAGAAGGACCTGGCAGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605941 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SART1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SART1 - squamous cell carcinoma antigen recognized by T-cells 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005146.4 | 751 | Silent Mutation | CTA,CTG | L,L 210 | NP_005137.1 | |
XM_011545344.1 | 751 | Silent Mutation | CTA,CTG | L,L 112 | XP_011543646.1 | |
XM_011545345.2 | 751 | Silent Mutation | CTA,CTG | L,L 52 | XP_011543647.1 |
TSGA10IP - testis specific 10 interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |