Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGCTGGTGGGGGTGGTGGCAGCG[A/C]CAGCCGAGTCGACACTGGCCACCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142410 | ||||||||||||||||||||
Literature Links: |
C12orf43 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C12orf43 - chromosome 12 open reading frame 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286191.1 | 552 | Missense Mutation | GGC,GTC | G,V 238 | NP_001273120.1 | |
NM_001286192.1 | 552 | Missense Mutation | GGC,GTC | G,V 208 | NP_001273121.1 | |
NM_001286195.1 | 552 | Missense Mutation | GGC,GTC | G,V 197 | NP_001273124.1 | |
NM_001286196.1 | 552 | Missense Mutation | GGC,GTC | G,V 196 | NP_001273125.1 | |
NM_001286197.1 | 552 | Missense Mutation | GGC,GTC | G,V 175 | NP_001273126.1 | |
NM_001286198.1 | 552 | Missense Mutation | GGC,GTC | G,V 166 | NP_001273127.1 | |
NM_022895.2 | 552 | Missense Mutation | GGC,GTC | G,V 207 | NP_075046.1 | |
XM_017019829.1 | 552 | Intron | XP_016875318.1 |
HNF1A - HNF1 homeobox A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |