Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAACGGATTCAGGAACGGGTGAAG[A/G]CATTCCCAAGTGCCTGTCCTTATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615341 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP4A22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP4A22 - cytochrome P450 family 4 subfamily A member 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010969.3 | Intron | NP_001010969.2 | ||||
NM_001308102.1 | Intron | NP_001295031.1 | ||||
XM_005270767.3 | Intron | XP_005270824.1 | ||||
XM_005270770.3 | Intron | XP_005270827.1 |