Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACGAGTATTTAGATATGTGGGAGA[C/T]TGTTCAAAAACCAGCAAATCCGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611995 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MANSC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MANSC4 - MANSC domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146221.1 | 396 | Silent Mutation | CAA,CAG | Q,Q 132 | NP_001139693.1 | |
XM_006719024.3 | 396 | Silent Mutation | CAA,CAG | Q,Q 87 | XP_006719087.1 | |
XM_011520542.2 | 396 | Silent Mutation | CAA,CAG | Q,Q 105 | XP_011518844.1 |
MRPS35 - mitochondrial ribosomal protein S35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |