Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGCACCTGCCCTCAGCCAGGGA[A/G]AGGAAAGAAACTGGAACTGCCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610012 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SULF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SULF1 - sulfatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128204.1 | Intron | NP_001121676.1 | ||||
NM_001128205.1 | Intron | NP_001121677.1 | ||||
NM_001128206.1 | Intron | NP_001121678.1 | ||||
NM_015170.2 | Intron | NP_055985.2 | ||||
XM_006716438.2 | Intron | XP_006716501.1 | ||||
XM_006716439.2 | Intron | XP_006716502.1 | ||||
XM_006716440.3 | Intron | XP_006716503.1 | ||||
XM_006716441.1 | Intron | XP_006716504.1 | ||||
XM_006716442.1 | Intron | XP_006716505.1 | ||||
XM_011517494.1 | Intron | XP_011515796.1 | ||||
XM_011517495.1 | Intron | XP_011515797.1 | ||||
XM_017013250.1 | Intron | XP_016868739.1 |