Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTTCAATCCCATCGGCCATTTC[A/C]ATCACCTTGTCTAAAAGGTCTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKN2AIPNL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDKN2AIPNL - CDKN2A interacting protein N-terminal like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080656.2 | 323 | Missense Mutation | ATG,ATT | M,I 90 | NP_542387.1 | |
XM_011543709.2 | 323 | Intron | XP_011542011.1 |
LOC105379186 - uncharacterized LOC105379186 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |