Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCAAAGAAACCGACCCTGATTA[C/T]GAGGATGTATCTGTGGCCCTTCCAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 613373 | |||||||||||||||||||||||
Literature Links: |
YEATS2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
YEATS2 - YEATS domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018023.4 | 151 | Silent Mutation | TAC,TAT | Y,Y 15 | NP_060493.3 | |
XM_005247597.2 | 151 | Silent Mutation | TAC,TAT | Y,Y 15 | XP_005247654.1 | |
XM_005247598.4 | 151 | Silent Mutation | TAC,TAT | Y,Y 15 | XP_005247655.1 | |
XM_006713688.3 | 151 | Silent Mutation | TAC,TAT | Y,Y 15 | XP_006713751.1 | |
XM_011512965.1 | 151 | Silent Mutation | TAC,TAT | Y,Y 15 | XP_011511267.1 | |
XM_011512966.1 | 151 | Silent Mutation | TAC,TAT | Y,Y 15 | XP_011511268.1 | |
XM_017006809.1 | 151 | Silent Mutation | TAC,TAT | Y,Y 15 | XP_016862298.1 | |
XM_017006810.1 | 151 | Silent Mutation | TAC,TAT | Y,Y 15 | XP_016862299.1 | |
XM_017006811.1 | 151 | Intron | XP_016862300.1 | |||
XM_017006812.1 | 151 | Silent Mutation | TAC,TAT | Y,Y 15 | XP_016862301.1 | |
XM_017006813.1 | 151 | UTR 5 | XP_016862302.1 | |||
XM_017006814.1 | 151 | UTR 5 | XP_016862303.1 |