Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGCCTCGGGAGCGCTGGCGACT[A/C]GTGCTGCGCTCTTGTCTCGGGAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114217 | ||||||||||||||||||||
Literature Links: |
CANX PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CANX - calnexin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024649.1 | 1020 | Intron | NP_001019820.1 | |||
NM_001746.3 | 1020 | Intron | NP_001737.1 | |||
XM_011534664.1 | 1020 | Intron | XP_011532966.1 | |||
XM_011534665.2 | 1020 | Intron | XP_011532967.1 |
CBY3 - chibby family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164444.1 | 1020 | Silent Mutation | ACG,ACT | T,T 37 | NP_001157916.1 | |
XM_017009726.1 | 1020 | Silent Mutation | ACG,ACT | T,T 36 | XP_016865215.1 |
LOC105377763 - uncharacterized LOC105377763 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |