Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAAAAAGGAATTCCCCACCCCAC[A/C]CGGCAGGGTCGGCCCAGGTAAGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601890 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLC4 - kinesin light chain 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTK7 - protein tyrosine kinase 7 (inactive) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270398.1 | 151 | Missense Mutation | CAC,CCC | H,P 29 | NP_001257327.1 | |
NM_002821.4 | 151 | Intron | NP_002812.2 | |||
NM_152880.3 | 151 | Intron | NP_690619.1 | |||
NM_152881.3 | 151 | Intron | NP_690620.1 | |||
NM_152882.3 | 151 | Intron | NP_690621.1 | |||
XM_011514765.2 | 151 | Missense Mutation | CAC,CCC | H,P 29 | XP_011513067.1 | |
XM_011514766.2 | 151 | Missense Mutation | CAC,CCC | H,P 29 | XP_011513068.1 |