Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACATGAGCCTCCTCCTCTCCTTCT[A/G]CCTGCTGGGGTTGCTTGTCAGTAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602040 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCAM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCAM2 - neural cell adhesion molecule 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004540.3 | 135 | Missense Mutation | TAC,TGC | Y,C 8 | NP_004531.2 | |
XM_011529575.2 | 135 | Intron | XP_011527877.1 | |||
XM_011529576.2 | 135 | UTR 5 | XP_011527878.1 | |||
XM_011529579.2 | 135 | Missense Mutation | TAC,TGC | Y,C 8 | XP_011527881.1 | |
XM_011529580.2 | 135 | Intron | XP_011527882.1 | |||
XM_011529581.2 | 135 | Intron | XP_011527883.1 | |||
XM_011529582.2 | 135 | Intron | XP_011527884.1 | |||
XM_011529585.2 | 135 | Intron | XP_011527887.1 | |||
XM_017028353.1 | 135 | UTR 5 | XP_016883842.1 | |||
XM_017028354.1 | 135 | Missense Mutation | TAC,TGC | Y,C 8 | XP_016883843.1 | |
XM_017028355.1 | 135 | Missense Mutation | TAC,TGC | Y,C 8 | XP_016883844.1 | |
XM_017028356.1 | 135 | UTR 5 | XP_016883845.1 | |||
XM_017028357.1 | 135 | UTR 5 | XP_016883846.1 | |||
XM_017028358.1 | 135 | Intron | XP_016883847.1 |