Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604699 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFRP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARFRP1 - ADP ribosylation factor related protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZGPAT - zinc finger CCCH-type and G-patch domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083113.1 | Intron | NP_001076582.1 | ||||
NM_001195653.1 | Intron | NP_001182582.1 | ||||
NM_001195654.1 | Intron | NP_001182583.1 | ||||
NM_032527.4 | Intron | NP_115916.3 | ||||
NM_181485.2 | Intron | NP_852150.2 |